Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IQCA1 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988756
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8756 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8756 Supplier Bioss Supplier No. BS9020R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene is a member of the ATPases Associated with diverse cellular Activities (AAA) superfamily. Members of this superfamily, found in all organisms, participate in a large number of cellular processes and contain the ATPase module consisting of an alpha-beta-alpha core domain and the Walker A and B motifs of the P-loop NTPases. Alternative splicing results in multiple transcript variants that encode different protein isoforms. [provided by RefSeq, Jul 2012].

Specifications

Antigen IQCA1
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene IQCA1
Gene Accession No. Q86XH1
Gene Alias 4930465P12Rik; DRC11; Dynein regulatory complex protein 11; dynein regulatory complex subunit 11; IQ and AAA domain-containing protein 1; IQ motif containing with AAA domain 1; IQCA; IQCA1
Gene Symbols IQCA1
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human IQCA1.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79781
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.