Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IQCB1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101645
Description
Antibody detects endogenous levels of total IQCB1.
This gene was first characterized as part of a cluster of genes located within the human major histocompatibility complex class III region. This gene encodes a nuclear protein that is cleaved by caspase 3 and is implicated in the control of apoptosis. In addition, the protein forms a complex with E1A binding protein p300 and is required for the acetylation of p53 in response to DNA damage. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
IQCB1 | |
Polyclonal | |
Unconjugated | |
Iqcb1 | |
6820449I09Rik; AV128382; IQ calmodulin-binding motif containing 1; IQ calmodulin-binding motif-containing protein 1; IQ motif containing B1; IQCB1; KIAA0036; nephrocystin 5; Nephrocystin-5; NPHP5; OK/SW-cl.85; p53 and DNA damage-regulated IQ motif protein; PIQ; SLSN5 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
9657 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q15051 | |
Iqcb1 | |
A synthesized peptide derived from human IQCB1(Accession Q15051), corresponding to amino acid residues C434-A484. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction