Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IQCB1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112923
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112923 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112923 Supplier Invitrogen™ Supplier No. PA5112923
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IQCB1 Polyclonal Antibody for Western Blot, IHC (P), ELISA

This gene was first characterized as part of a cluster of genes located within the human major histocompatibility complex class III region. This gene encodes a nuclear protein that is cleaved by caspase 3 and is implicated in the control of apoptosis. In addition, the protein forms a complex with E1A binding protein p300 and is required for the acetylation of p53 in response to DNA damage. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IQCB1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.03 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Iqcb1
Gene Accession No. Q15051, Q8BP00
Gene Alias 6820449I09Rik; AV128382; IQ calmodulin-binding motif containing 1; IQ calmodulin-binding motif-containing protein 1; IQ motif containing B1; IQCB1; KIAA0036; nephrocystin 5; Nephrocystin-5; NPHP5; OK/SW-cl.85; p53 and DNA damage-regulated IQ motif protein; PIQ; SLSN5
Gene Symbols Iqcb1
Host Species Rabbit
Immunogen Recombinant Human IQ calmodulin-binding motif-containing protein 1 protein (445-598AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 303915, 320299, 9657
Target Species Human, Mouse, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.