Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IQCB1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501570588
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-157-0588 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-157-0588 Supplier Bethyl Laboratories, Inc Supplier No. A305499A
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Mouse

The nephrocystin proteins comprise a family of five enzymes that commonly interact with p130Cas, proline-rich tyrosine kinases, calmodulin and tensin, indicating that these proteins may participate in a common signaling pathway. Localized to the outer segments and primary cilia of photoreceptor cells, nephrocystin-5 is complexed with RPGR (retinitis pigmentosa GTPase regulator) and interacts directly with calmodulin. Nephrocystin-5 is thought to participate with RPGR in a pathway of ciliary function in the kidey and retina. Mutations in the gene encoding nephrocystin-5 are the primary cause of Senior-Loken syndrome 5, a juvenile disorder characterized by defects in the waste filtering system of the kidney, as well as retinal degradation.

Specifications

Antigen IQCB1
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene IQCB1
Gene Accession No. Q15051
Gene Alias IQ calmodulin-binding motif-containing protein 1, Nephrocystin-5, p53 and DNA damage-regulated IQ motif protein, PIQ, IQCB1, KIAA0036, NPHP5, OK/SW-cl.85
Gene Symbols IQCB1
Host Species Rabbit
Immunogen Between 548 and 598
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9657
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.