Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IQCC Polyclonal Antibody

Catalog No. PIPA535902
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA535902 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA535902 Supplier Invitrogen™ Supplier No. PA535902
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

The IQCC (IQ Motif Containing C) gene encodes a protein that contains the IQ motif, which is known for its role in binding calmodulin and other proteins with EF-hand calcium-binding motifs. IQCC is located on chromosome 6 at position 6q16.1. The IQCC protein is involved in several cellular processes, including signaling pathways related to intracellular calcium levels and regulation of ion channels. Additionally, recent studies have indicated its potential involvement in cancer biology, where disruptions in its expression or function may contribute to tumorigenesis. The exact molecular mechanisms and pathways influenced by IQCC are still under investigation, and elucidating these aspects could uncover new therapeutic targets for various diseases, including cancer.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IQCC
Applications Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.01% thimerosal; pH 7
Gene IQCC
Gene Accession No. Q4KMZ1
Gene Alias BC026978; IQ domain-containing protein C; IQ motif containing C; Iqcc; RP4-622L5.6
Gene Symbols IQCC
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human IQCC. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55721
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.