Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IQCF2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA572886
Description
Specificity: IQCF2 antibody is human specic. IQCF2 antibody is predicted to not cross-react with IQCF1.

Specifications
IQCF2 | |
Polyclonal | |
Unconjugated | |
IQCF2 | |
IQ domain-containing protein F2; IQ motif containing F2; IQCF2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
389123 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
Q8IXL9 | |
IQCF2 | |
IQCF2 antibody for a 19 amino acid peptide near the amino terminus of human IQCF2. The immunogen is within the first 50 amino acids of IQCF2. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction