Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IQCG Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113357
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113357 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113357 Supplier Invitrogen™ Supplier No. PA5113357
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IQCG Polyclonal Antibody for Western Blot, ICC/IF, ELISA

IQCG is a protein coding gene. Component of the nexin-dynein regulatory complex (N-DRC), a key regulator of ciliary/flagellar motility which maintains the alignment and integrity of the distal axoneme and regulates microtubule sliding in motile axonemes. Binds calmodulin when cellular Ca(2+) levels are low and thereby contributes to the regulation of calcium and calmodulin-dependent protein kinase IV (CAMK4) activity; contributes to the regulation of CAMK4 signaling cascades. Required for normal axoneme assembly in sperm flagella, normal sperm tail formation and for male fertility.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IQCG
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene IQCG
Gene Accession No. Q80W32, Q9H095
Gene Alias 2400003L07Rik; CFAP122; DKFZp434B227; Drc9; Dynein regulatory complex protein 9; dynein regulatory complex subunit 9; esgd12d; G1-374-12; IQ domain-containing protein G; IQ motif containing G; Iqcg; repro1; RGD1311606; stubby12d
Gene Symbols IQCG
Host Species Rabbit
Immunogen Recombinant Human IQ domain-containing protein G protein (1-134AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 69707, 84223
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.