Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IQCG Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988758
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8758 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8758 Supplier Bioss Supplier No. BS9022R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IQCG is a protein coding gene. Component of the nexin-dynein regulatory complex (N-DRC), a key regulator of ciliary/flagellar motility which maintains the alignment and integrity of the distal axoneme and regulates microtubule sliding in motile axonemes. Binds calmodulin when cellular Ca(2+) levels are low and thereby contributes to the regulation of calcium and calmodulin-dependent protein kinase IV (CAMK4) activity; contributes to the regulation of CAMK4 signaling cascades. Required for normal axoneme assembly in sperm flagella, normal sperm tail formation and for male fertility.

Specifications

Antigen IQCG
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene IQCG
Gene Accession No. Q9H095
Gene Alias CFAP122; DKFZp434B227; DRC9; Dynein regulatory complex protein 9; dynein regulatory complex subunit 9; IQ domain-containing protein G; IQ motif containing G; IQCG
Gene Symbols IQCG
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human IQCG.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 84223
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.