Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IQCH Monoclonal Antibody (4H3)
GREENER_CHOICE

Catalog No. PIMA522514
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522514 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522514 Supplier Invitrogen™ Supplier No. MA522514
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: LSQLITDHLQ IQRWLFKMDS EFRGNGTAFC DIPSYLKCYK WVLKESSRYG LEDWRKKWAQ EPALVKISEE LAGILAQHAQ PVNEKRFPTW RKFLQTFLSQ.

May play a regulatory role in spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IQCH
Applications ELISA
Classification Monoclonal
Clone 4H3
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene IQCH
Gene Accession No. Q86VS3
Gene Alias IQ domain-containing protein H; IQ motif containing H; IQCH; NYDSP5; Testis development protein NYD-SP5
Gene Symbols IQCH
Host Species Mouse
Immunogen IQCH (NP_073621, 301 a.a. ~ 400 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64799
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.