Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IQCH Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559321
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559321 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559321 Supplier Invitrogen™ Supplier No. PA559321
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PLRALKSLWD YDFLIYDGVI DNTAPDFLAF KEHFSLAWGG IFSLLEHVEK FLRNYAIPEV KIKGNNLVAL LPEFELTNKL TRYDLLSVLE DPAHVQMLIN LPGQRYKGQD Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 60%.

May play a regulatory role in spermatogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IQCH
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IQCH
Gene Accession No. Q86VS3
Gene Alias IQ domain-containing protein H; IQ motif containing H; IQCH; NYDSP5; Testis development protein NYD-SP5
Gene Symbols IQCH
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IQCH. Recombinant protein control fragment (Product #RP-98037).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64799
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.