Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IQGAP1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP32144325UL
Description
IQGAP1 Polyclonal antibody specifically detects IQGAP1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
IQGAP1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin 1:200-1:1000 | |
HUMORFA01, IQ motif containing GTPase activating protein 1, KIAA0051ras GTPase-activating-like protein IQGAP1, p195RasGAP-like with IQ motifs, SAR1 | |
Produced in rabbits immunized with E. coli-derived Human IQGAP1 fragment | |
25 μg | |
Signal Transduction | |
8826 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Unconjugated | |
PBS, pH 7.0 | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction