Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IQGAP1 Mouse anti-Human, Clone: OTI7F5, lyophilized, TrueMAB™
SDP

Catalog No. 501669796
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-166-9796 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-166-9796 Supplier OriGene Supplier No. CF809928
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO

DISP2 is the second of three known homologs of the D. melanogaster protein Dispatched. It is a multi-transmembrane protein containing two PTCH/DISP domains and is thought to be involved in the release of lipid-anchored Hedgehog from producing cells. Hedgehog is a major player in signaling pathways during embryogenesis, tissue regeneration, and carcinogenesis and the DISP proteins have been implicated in these pathways. Recently, it has been shown that DISP2 is translationally regulated by the microRNA miR-214 in zebrafish. Expression of this miRNA decreased DISP2 promoter activity in vitro and its overexpression in zebrafish resulted in a phenotype identical to that observed by DISP2 mutants.

Specifications

Antigen IQGAP1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI7F5
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene IQGAP1
Gene Accession No. P46940
Gene Alias HUMORFA01, SAR1, p195
Gene Symbols IQGAP1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 464-609 of human IQGAP1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8826
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.