Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IQGAP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596385
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596385 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596385 Supplier Invitrogen™ Supplier No. PA596385
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ELMKLKAQAH AENNEFITWN DIQACVDHVN LVVQEEHERI LAIGLINEAL DEGDAQKTLQ ALQIPAAKLE GVLAEVAQHY QDTLIRAKRE KAQEIQDESA VLWLDEIQGG IWQSNKDTQE AQKFALGIFA INEAVESGDV GKTLSALRSP DVGLYGVIPE CGETYHSDLA EAKKKKLAVG DNNSKWVKHW VKGGYYYYHN LETQEGGWDE PPNFVQNSMQ LSREEIQSSI SGVTAAYNRE QLWLANEGLI T; Positive Samples: A-549, 293T, HepG2, HT-29, HeLa; Cellular Location: Cell membrane.

This gene encodes a member of the IQGAP family. The protein contains four IQ domains, one calponin homology domain, one Ras-GAP domain and one WW domain. It interacts with components of the cytoskeleton, with cell adhesion molecules, and with several signaling molecules to regulate cell morphology and motility. Expression of the protein is upregulated by gene amplification in two gastric cancer cell lines.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IQGAP1
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1.07 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene IQGAP1
Gene Accession No. P46940, Q9JKF1
Gene Alias AA682088; Cdc42-Rac1 effector protein; D7Ertd237e; D7Ertd257e; DISP2; DISPB; HUMORFA01; IQ motif containing GTPase activating protein 1; IQGAP1; KIAA0051; mKIAA0051; p195; Ras GTPase-activating-like protein IQGAP1; RasGAP-like with IQ motifs; SAR1
Gene Symbols IQGAP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 500-750 of human IQGAP1 (NP_0038611).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29875, 361598, 8826
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.