Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IQGAP1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501561394
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-1394 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-1394 Supplier Bethyl Laboratories, Inc Supplier No. A301950AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.

DISP2 is the second of three known homologs of the D. melanogaster protein Dispatched. It is a multi-transmembrane protein containing two PTCH/DISP domains and is thought to be involved in the release of lipid-anchored Hedgehog from producing cells. Hedgehog is a major player in signaling pathways during embryogenesis, tissue regeneration, and carcinogenesis and the DISP proteins have been implicated in these pathways. Recently, it has been shown that DISP2 is translationally regulated by the microRNA miR-214 in zebrafish. Expression of this miRNA decreased DISP2 promoter activity in vitro and its overexpression in zebrafish resulted in a phenotype identical to that observed by DISP2 mutants.

Specifications

Antigen IQGAP1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene IQGAP1
Gene Accession No. P46940
Gene Alias IQGAP1; IQ motif containing GTPase activating protein 1; HUMORFA01; KIAA0051; SAR1; p195; RasGAP-like with IQ motifs
Gene Symbols IQGAP1
Host Species Rabbit
Immunogen Between 1 and 50
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8826
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.