Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IQGAP1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA578262
Description
Positive Control: 293T, A431, HeLa, HepG2 Predicted Reactivity: Mouse (87%), Rat (88%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a member of the IQGAP family. The protein contains four IQ domains, one calponin homology domain, one Ras-GAP domain and one WW domain. It interacts with components of the cytoskeleton, with cell adhesion molecules, and with several signaling molecules to regulate cell morphology and motility. Expression of the protein is upregulated by gene amplification in two gastric cancer cell lines.
Specifications
IQGAP1 | |
Polyclonal | |
Unconjugated | |
IQGAP1 | |
AA682088; Cdc42-Rac1 effector protein; D7Ertd237e; D7Ertd257e; DISP2; DISPB; HUMORFA01; IQ motif containing GTPase activating protein 1; IQGAP1; KIAA0051; mKIAA0051; p195; Ras GTPase-activating-like protein IQGAP1; RasGAP-like with IQ motifs; SAR1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8826 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1.16 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
P46940 | |
IQGAP1 | |
Recombinant protein encompassing a sequence within the center region of human IQGAP1. The exact sequence is proprietary. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction