Learn More
IQGAP1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A301950A

Description
The recommended shelf life for this product is 1 year from date of receipt. Application Note: For IHC, epitope retrieval with citrate buffer pH 6.0 is recommended for FFPE tissue sections.
DISP2 is the second of three known homologs of the D. melanogaster protein Dispatched. It is a multi-transmembrane protein containing two PTCH/DISP domains and is thought to be involved in the release of lipid-anchored Hedgehog from producing cells. Hedgehog is a major player in signaling pathways during embryogenesis, tissue regeneration, and carcinogenesis and the DISP proteins have been implicated in these pathways. Recently, it has been shown that DISP2 is translationally regulated by the microRNA miR-214 in zebrafish. Expression of this miRNA decreased DISP2 promoter activity in vitro and its overexpression in zebrafish resulted in a phenotype identical to that observed by DISP2 mutants.Specifications
IQGAP1 | |
Polyclonal | |
Unconjugated | |
IQGAP1 | |
IQGAP1; IQ motif containing GTPase activating protein 1; HUMORFA01; KIAA0051; SAR1; p195; RasGAP-like with IQ motifs | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
8826 | |
4° C | |
Liquid |
Immunohistochemistry, Immunoprecipitation, Western Blot | |
0.2 mg/ml | |
TBS with 0.1% BSA and 0.09% sodium azide | |
P46940 | |
IQGAP1 | |
Between 1 and 50 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.