Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IQGAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5120356
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5120356 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5120356 Supplier Invitrogen™ Supplier No. PA5120356
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Jurkat, Mouse liver. Immunogen sequence: QQEVDHATDM VSRAMIDSRT PEEMKHSQSM IEDAQLPLEQ KKRKIQRNLR TLEQTGHVSS ENKYQDILNE IAKDIRNQRI YRKLRKAELA KLQQTLNALN KKAAFYEEQI N.

Binds to activated CDC42 and RAC1 but does not seem to stimulate their GTPase activity. Associates with calmodulin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IQGAP2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.71 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene IQGAP2
Gene Accession No. Q13576, Q3UQ44
Gene Alias 4933417J23Rik; A630053O10; AI788777; IQ motif containing GTPase activating protein 2; Iqgap2; Ras GTPase-activating-like protein IQGAP2
Gene Symbols IQGAP2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1330-1440 of human IQGAP2 (NP_006624.2).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100360623, 10788, 544963
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.