Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IQSEC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558268
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558268 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558268 Supplier Invitrogen™ Supplier No. PA558268
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GSLKRQSAYE RSLGGQQGSP KHGPHSGAPK SLPREEPELR PRPPRPLDSH LAINGSANRQ SKSESDYSDG DNDSINSTSN S Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 94%.

ADP ribosylation factors (Arfs) are small GTP-binding proteins known for their role in vesicular transport, where they nucleate the assembly of coat protein complexes at sites of carrier vesicle formation. IQSEC1 (GEP100/BRAG2), a guanine nucleotide exchange protein for ARF6, belongs to the BRAG family and contains one IQ domain, one PH domain and one SEC7 domain. As a known activator of ARF6, IQSEC1 regulates cell surface levels of Beta1 integrin and its knockdown blocked tumour metastasis. IQSEC1 links EGFR signaling to ARF6 activation and may contribute to the cancer metastasis and malignancy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IQSEC1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene IQSEC1
Gene Accession No. Q6DN90
Gene Alias ADP-ribosylation factors guanine nucleotide-exchange protein 100; ADP-ribosylation factors guanine nucleotide-exchange protein 2; ARFGEP100; ARF-GEP100; AW561907; BC026481; BRAG2; brefeldin A-resistant ARF-GEF2; brefeldin-resistant Arf-GEF 2 protein; cI-43; D6Ertd349e; GEP100; IQ motif and Sec7 domain 1; IQ motif and SEC7 domain-containing protein 1; Iqsec1; Kiaa0763
Gene Symbols IQSEC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human IQSEC1. Recombinant protein control fragment (Product #RP-96356).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9922
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.