Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IQSEC2 Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS13645R

Description
This gene encodes a guanine nucleotide exchange factor for the ARF family of small GTP-binding proteins. The encoded protein is a component of the postsynaptic density at excitatory synapses, and may play a critical role in cytoskeletal and synaptic organization through the activation of selected ARF substrates including ARF1 and ARF6. Mutations in this gene have been implicated in nonsyndromic X-linked mental retardation. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene.Specifications
IQSEC2 | |
Polyclonal | |
Unconjugated | |
IQSEC2 | |
BRAG1; brefeldin A resistant Arf-guanine nucleotide exchange factor 1; brefeldin A resistant Arf-guanine nucleotide exchange factor 1b; brefeldin A resistant Arf-guanine nucleotide exchange factor 1c; IQ motif and Sec7 domain 2; IQ motif and SEC7 domain-containing protein 2; IQSEC2; Kiaa0522; mKIAA0522; MRX1; MRX78; RGD1562109 | |
Rabbit | |
Protein A | |
RUO | |
23096 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
Q5JU85 | |
IQSEC2 | |
KLH conjugated synthetic peptide derived from human IQSEC2. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction