Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IQSEC3 Rabbit anti-Human, Mouse, Rat, Polyclonal, FabGennix

Catalog No. 502207486
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-7486 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-7486 Supplier Fabgennix Inc Supplier No. IQSEC3301AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

IQSEC3 Polyclonal antibody specifically detects IQSEC3 in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen IQSEC3
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene IQSEC3
Gene Accession No. Q3TES0, Q76M68, Q9UPP2
Gene Alias IQ motif and Sec7 domain 3; IQ motif and SEC7 domain-containing protein 3; IQSEC3; KIAA1110
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 1100-1150 on human IQ motif and SEC7 domain-containing protein 3.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 243621, 404781, 440073
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.