Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals IR-125, pure, laser grade

Catalog No. AC412540020
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC412540020 2 g Glass bottle
AC412541000 100 mg Glass bottle
AC412545000 500 mg Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC412540020 Supplier Thermo Scientific Chemicals Supplier No. 412540020
Add to Cart

CAS: 3599-32-4 | C43H47N2NaO6S2 | 774.967 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3599-32-4
Molecular Formula C43H47N2NaO6S2
Molecular Weight (g/mol) 774.967
MDL Number MFCD00013078
InChI Key MOFVSTNWEDAEEK-UHFFFAOYSA-M
Synonym Indocyanine Green
PubChem CID 132274068
ChEBI CHEBI:31696
IUPAC Name sodium;4-[(2E)-2-[(2E,4E,6Z)-7-[1,1-dimethyl-3-(4-sulfonatobutyl)benzo[e]indol-3-ium-2-yl]hepta-2,4,6-trienylidene]-1,1-dimethylbenzo[e]indol-3-yl]butane-1-sulfonate
SMILES CC1(C(=[N+](C2=C1C3=CC=CC=C3C=C2)CCCCS(=O)(=O)[O-])C=CC=CC=CC=C4C(C5=C(N4CCCCS(=O)(=O)[O-])C=CC6=CC=CC=C65)(C)C)C.[Na+]

Specifications

Melting Point 235.0°C
Merck Index 15, 5002
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in methanol, dmso, acetonitrile
Infrared Spectrum Authentic
Lambda Max 784nm (in methanol)
Formula Weight 774.97
Grade Pure
Molar Absorption Coefficient 237500L/(mole.cm) (dried sample)
Packaging Glass bottle
Color Brown to Green
Quantity 2 g
Physical Form Crystalline Powder or Crystals
Chemical Name or Material IR-125
Show More Show Less
Hazard Category HYGROSCOPIC
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 222-751-5
RTECSNumber DE3150000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.