Learn More
Invitrogen™ IRAK-M Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120919
Description
Positive test controls include: THP-1, NCI-H125, Rat liver. Immunogen sequence: VLTGCRVVLD DPKHIQLRDL LRELMEKRGL DSCLSFLDKK VPPCPRNFSA KLFCLAGRCA ATRAKLRPSM DEVLNTLEST QASLYFAEDP PTSLKSFRCP SPLFLENVPS IPVEDDESQN NNLLPSDEGL RIDRMTQKTP FECSQSEVMF LSLDKKPESK RNEEACNMPS SSCEESWFPK YIVPSQDLRP YKVNIDPSSE APGHSCRSRP VESSCSSKFS WDEYEQYKKE.
IRAK3 serine/threonine kinase or interleukin-1 receptor-associated kinase 3 is a member of the interleukin-1 receptor-associated kinase protein family which are essential components of the Toll/IL-R immune signal transduction pathways. IRAK3 is primarily expressed in monocytes and macrophages and functions as a negative regulator of Toll-like receptor signaling. IRAK3 mediates critical aspects of innate immunity that result in an immunocompromised state during sepsis. Mutations in IRAK3 are associated with a susceptibility to asthma.
Specifications
IRAK-M | |
Polyclonal | |
Unconjugated | |
IRAK3 | |
4833428C18Rik; AI563835; ASRT5; FLJ13601; IL-1 receptor-associated kinase M; interleukin 1 receptor associated kinase 3; interleukin-1 receptor-associated kinase 3; IRAK3; IRAK-3; IRAKM; IRAK-M | |
Rabbit | |
Affinity Chromatography | |
RUO | |
11213, 314870, 73914 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
3.39 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K4B2, Q9Y616 | |
IRAK3 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 367-596 of human IRAK3 (NP_009130.2). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.