Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IRAK-M Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA586500
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA586500 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA586500 Supplier Invitrogen™ Supplier No. PA586500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Purity is > 95% by SDS-PAGE.

IRAK3 serine/threonine kinase or interleukin-1 receptor-associated kinase 3 is a member of the interleukin-1 receptor-associated kinase protein family which are essential components of the Toll/IL-R immune signal transduction pathways. IRAK3 is primarily expressed in monocytes and macrophages and functions as a negative regulator of Toll-like receptor signaling. IRAK3 mediates critical aspects of innate immunity that result in an immunocompromised state during sepsis. Mutations in IRAK3 are associated with a susceptibility to asthma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRAK-M
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene IRAK3
Gene Accession No. Q9Y616
Gene Alias 4833428C18Rik; AI563835; ASRT5; FLJ13601; IL-1 receptor-associated kinase M; interleukin 1 receptor associated kinase 3; interleukin-1 receptor-associated kinase 3; IRAK3; IRAK-3; IRAKM; IRAK-M
Gene Symbols IRAK3
Host Species Rabbit
Immunogen Synthetic peptide, corresponding to amino acids 500-540 of Human IRAK-M.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11213
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.