Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRAK1 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP177068
Description
IRAK1 Polyclonal specifically detects IRAK1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.Specifications
IRAK1 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 1, IRAK-1, IRAKPelle homolog, pelle | |
Rabbit | |
77 kDa | |
0.1 mg | |
Protein Kinase | |
3654 | |
Human, Mouse, Rat | |
IgG |
Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, Immunoprecipitation | |
1 mg/mL | |
Western Blot 1-4 ug/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 10-20 ug/mL, Immunoprecipitation 1:10-1:500 | |
P51617 | |
IRAK1 | |
Antibody was raised against a peptide corresponding to 13 amino acids near the carboxy terminus of human IRAK. The immunogen is located within the last 50 amino acids of IRAK. Amino Acid Squence: DRQGPEESDEFQS | |
Peptide affinity purified | |
RUO | |
Primary | |
At least four isoforms of IRAK1 are known to exist; this antibody will detect all four isoforms. IRAK1 antibody is predicted to not cross-react with other members of the IRAK protein family. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction