Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRAK1 Antibody (SR2210), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP321577100UL
Description
IRAK1 Monoclonal antibody specifically detects IRAK1 in Human samples. It is validated for Western Blot, Immunofluorescence, ImmunoprecipitationSpecifications
IRAK1 | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot, Immunofluorescence, Immunoprecipitation | |
SR2210 | |
Western Blot 1:500-1:1000, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunoprecipitation 1:50 | |
EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 1, IRAK-1, IRAKPelle homolog, pelle | |
A synthesized peptide derived from human IRAK1 (Uniprot #: P51617) | |
100 μg | |
Protein Kinase | |
3654 | |
Store at -20 degrees C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction