Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IRAK1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5141040
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5141040 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5141040 Supplier Invitrogen™ Supplier No. PA5141040
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

IRAK1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)

IL-1 receptor associated kinases (IRAKs) are serine/threonine kinases involved in signaling via Toll/IL-1 pathways. It is rapidly recruited by MYD88 to the receptor-signaling complex upon TLR activation. IRAK's association with MYD88 leads to IRAK1 phosphorylation by IRAK4 and subsequent autophosphorylation and kinase activation. Alternatively, IRAK1 can phosphorylate TIRAP to promote its ubiquitination and subsequent degradation. IRAK1 also phosphorylates the interferon regulatory factor 7 (IRF7) to induce its activation and translocation to the nucleus. This results in the transcriptional activiation of type I IFN genes, which drive the cell in an antiviral state.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRAK1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Irak1
Gene Accession No. P51617, Q62406
Gene Alias AA408924; Il1rak; interleukin 1 receptor associated kinase 1; interleukin-1 receptor-associated kinase 1; interleukin-1 receptor-associated kinase-1; IRAK; Irak1; IRAK-1; IRAK1-S; mPLK; pelle; Pelle homolog; pelle-like protein kinase; Plpk; RGD1563841
Gene Symbols Irak1
Host Species Rabbit
Immunogen Recombinant protein within human IRAK-1 aa 1-250.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16179, 3654
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.