Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRAK1 Rabbit anti-Human, Clone: 1H5U0, Novus Biologicals™
SDP

Catalog No. p-7247882 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB167922 20 μg
NB167921 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB167922 Supplier Novus Biologicals Supplier No. NBP31641520UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

IRAK1 Monoclonal antibody specifically detects IRAK1 in Human samples. It is validated for Western Blot

Specifications

Antigen IRAK1
Applications Western Blot
Classification Monoclonal
Clone 1H5U0
Conjugate Unconjugated
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 1, IRAK-1, IRAKPelle homolog, pelle
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 215-499 of human IRAK1 (P51617).
Purification Method Affinity purified
Quantity 20 μg
Regulatory Status RUO
Research Discipline Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 3654
Target Species Human
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.