Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRAK1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 104782AP150UL

Description
Interleukin-1 receptor-associated kinase 1 (IRAK1) is a serine/threonine-protein kinase that plays a critical role in initiating innate immune response against foreign pathogens. IRAK is associated with Toll-like receptor (TLR) and IL-1R signaling pathways. It is rapidly recruited by MYD88 to the receptor-signaling complex upon TLR activation. IRAK′s association with MYD88 leads to IRAK1 phosphorylation by IRAK4 and subsequent autophosphorylation and kinase activation. Alternatively, IRAK1 can phosphorylate TIRAP to promote its ubiquitination and subsequent degradation. IRAK1 also phosphorylates the interferon regulatory factor 7 (IRF7) to induce its activation and translocation to the nucleus. This results in the transcriptional activiation of type I IFN genes, which drive the cell in an antiviral state.Specifications
IRAK1 | |
Polyclonal | |
Unconjugated | |
IRAK1 | |
IRAK, IRAK 1, IRAK1, pelle | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
16179, 363520, 3654 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.23 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P51617, Q62406 | |
Irak1 | |
IRAK1 Fusion Protein Ag0728 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction