Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRAK1BP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5107149
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5107149 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5107149 Supplier Thermo Scientific Supplier No. PA5107149
Add to Cart
Add to Cart

IRAK1BP1 Polyclonal Antibody

IRAK1BP1 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen IRAK1BP1
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
Gene IRAK1BP1
Gene Accession No. Q9ESJ7
Gene Alias 4921528N06Rik; Aabp3; ActA binding protein 3; Acta-binding protein 70; AI851240; Aip70; interleukin 1 receptor associated kinase 1 binding protein 1; interleukin-1 receptor-associated kinase 1 binding protein 1; interleukin-1 receptor-associated kinase 1-binding protein 1; IRAK1-binding protein 1; Irak1bp1; MGC138458; MGC138460; PLK-interacting protein; signaling molecule that associates with the mouse pelle-like kinase; Simpl
Gene Symbols IRAK1BP1
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal region of Mouse Irak1bp1.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65099
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.