Learn More
Invitrogen™ IRAK1BP1 Polyclonal Antibody

Description
The antibody detects endogenous levels of total IRAK1BP1 protein.
Specifications
Specifications
| Antigen | IRAK1BP1 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Concentration | 0.3 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | IRAK1BP1 |
| Gene Accession No. | Q5VVH5, Q9ESJ7 |
| Gene Alias | 4921528N06Rik; Aabp3; ActA binding protein 3; Acta-binding protein 70; AI851240; Aip70; interleukin 1 receptor associated kinase 1 binding protein 1; interleukin-1 receptor-associated kinase 1 binding protein 1; interleukin-1 receptor-associated kinase 1-binding protein 1; IRAK1-binding protein 1; Irak1bp1; MGC138458; MGC138460; PLK-interacting protein; signaling molecule that associates with the mouse pelle-like kinase; Simpl |
| Gene Symbols | IRAK1BP1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.