Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IRAK1BP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578635
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578635 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578635 Supplier Invitrogen™ Supplier No. PA578635
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: Target recombinant protein Predicted Reactivity: Rhesus Monkey (97%), Bovine (88%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

SIMPL, also known as IRAK1BP1 (interleukin-1 receptor-associated kinase 1-binding protein 1), is a 260 amino acid protein belonging to the IRAK1BP1 family. In addition to containing an intrinsically disordered region which associates with HSPA1 and HSPA8, SIMPL also interacts directly with IRAK1 and promotes RELA transcriptional activity. SIMPL is a major component of the IRAK1-dependent TNFRSF1A signaling pathway in which TNF-alpha induces NF-kappa-B gene transcription, leading to an inhibition of proinflammatory cytokines. Inactivation of SIMPL inhibits tumor necrosis factor receptor type I-induced NF-kappaB activity. Localizing to the nucleus and cytoplasm, SIMPL requires the phosphorylation of Ser-56 and/or Ser-62 to be fully active.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRAK1BP1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 10% glycerol and 0.01% thimerosal; pH 7
Gene IRAK1BP1
Gene Accession No. Q5VVH5, Q9ESJ7
Gene Alias 4921528N06Rik; Aabp3; ActA binding protein 3; Acta-binding protein 70; AI851240; Aip70; interleukin 1 receptor associated kinase 1 binding protein 1; interleukin-1 receptor-associated kinase 1 binding protein 1; interleukin-1 receptor-associated kinase 1-binding protein 1; IRAK1-binding protein 1; Irak1bp1; MGC138458; MGC138460; PLK-interacting protein; signaling molecule that associates with the mouse pelle-like kinase; Simpl
Gene Symbols IRAK1BP1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human IRAK1BP1. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 134728, 300862, 65099
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.