Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRAK2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | IRAK2 |
---|---|
Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50 - 1:200 |
Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
IRAK2 Polyclonal specifically detects IRAK2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
IRAK2 | |
Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
O43187 | |
3656 | |
This antibody was developed against a recombinant protein corresponding to amino acids: AFLQPPEEDAPHSLRSDLPTSSDSKDFSTSIPKQEKLLSLAGDSLFWSEADVVQATDDFNQNRKISQGTFADVYRGHRHGKPFVFKKL | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
Polyclonal | |
Rabbit | |
Immunology, Innate Immunity, Protein Kinase, Signal Transduction | |
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
interleukin-1 receptor associated kinase-2, interleukin-1 receptor-associated kinase 2, interleukin-1 receptor-associated kinase-like 2, IRAK-2, MGC150550 | |
IRAK2 | |
IgG | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title