Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human IRAK2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB6690SP
This item is not returnable.
View return policy
Description
IRAK2 Monoclonal specifically detects IRAK2 in Human samples. It is validated for Western Blot.Specifications
IRAK2 | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL | |
O43187 | |
IRAK2 | |
E. coli-derived recombinant human IRAK2 Ala2-Asn590 Accession # O43187 | |
25 μg | |
Primary | |
Detects human IRAK2 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) IRAK1, 3, or 4 is observed. In Western blots, less than 25% cross-reactivity with rhIRAK1, 3, and 4 is observed. | |
Human | |
Purified |
Western Blot | |
600848 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
interleukin-1 receptor associated kinase-2, interleukin-1 receptor-associated kinase 2, interleukin-1 receptor-associated kinase-like 2, IRAK-2, MGC150550 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
3656 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG3 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction