Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRAK3 Rabbit anti-Human, Unconjugated, Clone: FDG-9, Abnova™
Rabbit Monoclonal Antibody.
Supplier: Abnova Corporation MAB20398
Description
IRAK3 Monoclonal antibody specifically detects IRAK3 in Human samples. It is validated for IP, WB-Ce.Specifications
IRAK3 | |
Monoclonal | |
Unconjugated | |
IRAK3 | |
Rabbit | |
100 μg | |
11213 | |
Store at -20°C for one year. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for a longer time. Avoid repeated freezing and thawing. | |
IgG |
Immunoprecipitation, Western Blot | |
FDG-9 | |
interleukin-1 receptor-associated kinase 3 | |
ASRT5, FLJ13601, IRAK-M, IRAKM | |
A synthetic peptide corresponding to human IRAK3 | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction