Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRAK4 Antibody (413613), PerCP, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals FAB39191C
Description
IRAK4 Monoclonal antibody specifically detects IRAK4 in Human samples. It is validated for Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOFSpecifications
IRAK4 | |
Monoclonal | |
PerCP | |
PBS | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
413613 | |
Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready | |
EC 2.7.11, EC 2.7.11.1, interleukin-1 receptor-associated kinase 4, IPD1, IRAK-4, NY-REN-64, REN64, Renal carcinoma antigen NY-REN-64 | |
E. coli-derived recombinant human IRAK4, Met1-Ser460, Accession # Q9NWZ3 | |
0.1 mL | |
Cytokine Research, Immunology, Innate Immunity, Protein Kinase | |
51135 | |
Store at 4°C in the dark. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction