Learn More
Promega IRAK4 Kinase Enzyme System, 10microg
Catalog No. PRV2621 Shop All Promega Corporation Products
Encompass
Change view
Click to view available options
Quantity:
10 μg
Catalog No. Quantity
PRV2621 10 μg
1 options
Catalog No. PRV2621 Supplier Promega Supplier No. V2621
Recombinant full-length human IRAK4 was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag. Interleukin-1 receptor-associated kinase 4 is an important mediator in the signal transduction of Toll-like receptor and IL1R family members.
  • Easily Screen and Profile IRAK4 Kinase Inhibitors
  • Includes kinase, substrate and reaction buffer
  • Use with ADP−Glo™ Assay for bioluminescent detection of kinase activity
Specifications
Quantity 10 μg
Content And Storage <-65°C