Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ IRAK4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA517531
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA517531 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA517531 Supplier Invitrogen™ Supplier No. PA517531
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

It is not recommended to aliquot this antibody.

IL-1 receptor associated kinases (IRAKs) are serine/threonine kinases involved in signaling via Toll/IL-1 pathways. Overexpression of IRAK-4 activates NF-kappa-B and MAPK pathways. IRAK-4 interacts with and phosphorylates IRAK-1. IRAK-4-deficient animals are completely resistant to the challenge with LPS. Animals and humans lacking IRAK-4 are impaired in their responses to viral and bacterial challenges. Members in IRAK/Pelle family play a central role in IL-1R/TLR mediated inflammatory responses and in innate immunity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRAK4
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 300 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene IRAK4
Gene Accession No. Q8R4K2, Q9NWZ3
Gene Alias 8430405M07Rik; 9330209D03Rik; interleukin 1 receptor associated kinase 4; interleukin-1; interleukin-1 receptor-associated kinase 4; IPD1; Irak4; IRAK-4; NY-REN-64; REN64; renal carcinoma antigen NY-REN-64
Gene Symbols IRAK4
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues surrounding lysine 41 of human IRAK4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 266632, 300177, 51135
Target Species Human, Mouse, Monkey, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.