Learn More
Invitrogen™ IRAK4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ 365500

Description
IRAK4 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
IL-1 receptor associated kinases (IRAKs) are serine/threonine kinases involved in signaling via Toll/IL-1 pathways. Overexpression of IRAK-4 activates NF-kappa-B and MAPK pathways. IRAK-4 interacts with and phosphorylates IRAK-1. IRAK-4-deficient animals are completely resistant to the challenge with LPS. Animals and humans lacking IRAK-4 are impaired in their responses to viral and bacterial challenges. Members in IRAK/Pelle family play a central role in IL-1R/TLR mediated inflammatory responses and in innate immunity.
Specifications
IRAK4 | |
Polyclonal | |
Unconjugated | |
IRAK4 | |
8430405M07Rik; 9330209D03Rik; interleukin 1 receptor associated kinase 4; interleukin-1; interleukin-1 receptor-associated kinase 4; IPD1; Irak4; IRAK-4; NY-REN-64; REN64; renal carcinoma antigen NY-REN-64 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
266632, 51135 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
0.25 mg/mL | |
PBS with 0.1% sodium azide; pH 7.4 | |
Q8R4K2, Q9NWZ3 | |
IRAK4 | |
Synthetic peptide derived from the N-terminal region of the human IRAK4 (IL-1 receptor associated kinase 4) protein. | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.