Learn More
Description
Application: Fluorescence microscopy.
Properties and Photophysical Data: Excitation and emission maxima (λ) are 778 nm and 794 nm, respectively.
It is recommended to dissolve this dye in water or DMSO (10 - 20 mg/mL)
Specifications
Specifications
| CAS | 1373928-39-2 |
| Quantity | 500 μg |
| Target | Fluorescent Dyes |
| Molecular Formula | C64H65N4Na3O15S4 |
| Purity | 0.8 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
