Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRE1 alpha Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 37 publications

$200.40 - $549.92

Specifications

Antigen IRE1 alpha
Concentration 1 mg/mL
Dilution Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:250 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:250, Immunohistochemistry-Paraffin 1:250 - 1:500, Knockout Validated reported in scientific literature (PMID 31159306), KnockDown Validated
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB1002324SS
SDP
View Documents
Novus Biologicals
NB1002324SS
0.025 mL
Each for $200.40
Only null left
Add to Cart
 
NB1002324
SDP
View Documents
Novus Biologicals
NB1002324
0.1 mL
Each for $549.92
Only null left
Add to Cart
 
Description

Description

IRE1 alpha Polyclonal specifically detects IRE1 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications

Specifications

IRE1 alpha
Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:250 - 1:500, Immunocytochemistry/Immunofluorescence 1:100 - 1:250, Immunohistochemistry-Paraffin 1:250 - 1:500, Knockout Validated reported in scientific literature (PMID 31159306), KnockDown Validated
Polyclonal
Rabbit
Apoptosis, Cell Biology, Diabetes Research, Hypoxia, Lipid and Metabolism, Neuroscience, Signal Transduction, Unfolded Protein Response, Unfolded Protein Response (UPR)
Tris-Glycine and 0.15M NaCl with 0.05% Sodium Azide
EC 2.7.11, endoplasmic reticulum to nucleus signaling 1, Endoplasmic reticulum-to-nucleus signaling 1, ER to nucleus signalling 1, FLJ30999, hIRE1p, inositol-requiring 1, Inositol-requiring protein 1, IRE 1a, IRE1a, Ire1-alpha, IRE1MGC163279, IRE1P, MGC163277, protein kinase/endoribonuclease, serine/threonine-protein kinase/endoribonuclease IRE1
ERN1
IgG
Affinity Purified
This antibody detects the levels of endogenous total IRE1 Alpha protein.
1 mg/mL
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human, Mouse, Rat
O75460
2081
This IRE1 alpha antibody was raised against a synthetic peptide within the human IRE1 alpha protein (within residues 700-800). [Swiss-Prot #O75460]
Primary
Store at -20C. Avoid freeze-thaw cycles.
110 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.