Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRE1 alpha Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00 - $610.00
Specifications
Antigen | IRE1 alpha |
---|---|
Dilution | Immunohistochemistry-Paraffin 1:200 - 1:500 |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
IRE1 alpha Polyclonal specifically detects IRE1 alpha in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
IRE1 alpha | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
RUO | |
Human | |
EC 2.7.11, endoplasmic reticulum to nucleus signaling 1, Endoplasmic reticulum-to-nucleus signaling 1, ER to nucleus signalling 1, FLJ30999, hIRE1p, inositol-requiring 1, Inositol-requiring protein 1, IRE 1a, IRE1a, Ire1-alpha, IRE1MGC163279, IRE1P, MGC163277, protein kinase/endoribonuclease, serine/threonine-protein kinase/endoribonuclease IRE1 | |
ERN1 | |
IgG | |
Affinity Purified | |
110 kDa |
Immunohistochemistry-Paraffin 1:200 - 1:500 | |
Polyclonal | |
Rabbit | |
Apoptosis, Cell Biology, Diabetes Research, Hypoxia, Lipid and Metabolism, Neuroscience, Signal Transduction, Unfolded Protein Response | |
PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide | |
2081 | |
This IRE1 alpha antibody was developed against a Recombinant Protein corresponding to amino acids: VTIGDKGECVITPSTDVKFDPGLKSKNKLNYLRNYWLLIGHHETPLSASTKMLERFPNNLPKHRENVIPADSEKKSFEEVINLVDQTSENAPTTVSRDVEEKPAHAPARPEAPVDSMLKDMATIILSTFLLIGWVA | |
Primary | |
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title