Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRE1 alpha Recombinant Rabbit Monoclonal Antibody (142)
GREENER_CHOICE

Catalog No. PIMA530533
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530533 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530533 Supplier Thermo Scientific Supplier No. MA530533
Add to Cart
Add to Cart

IRE1 alpha Recombinant Rabbit Monoclonal Antibody (142)

IRE1 alpha Recombinant Monoclonal Antibody for ELISA

TRUSTED_SUSTAINABILITY

Specifications

Antigen IRE1 alpha
Applications ELISA
Classification Recombinant Monoclonal
Clone 142
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ERN1
Gene Accession No. O75460
Gene Alias 9030414B18Rik; AI225830; C85377; endoplasmic reticulum (ER) to nucleus signalling 1; endoplasmic reticulum to nucleus signaling 1; endoplasmic reticulum to nucleus signalling 1; endoplasmic reticulum-to-nucleus signaling 1; Endoribonuclease; ER to nucleus signalling 1; ERN1; FLJ30999; hIRE1p; inositol-requiring 1; inositol-requiring 1 alpha; inositol-requiring enzyme 1; inositol-requiring protein 1; IRE1; IRE1a; IRE1a antibody; Ire1alpha; Ire1-alpha; Ire1p; IRE1p antibody; MGC163277; MGC163279; protein kinase/endoribonuclease; RGD1559716; Serine/threonine-protein kinase; Serine/threonine-protein kinase/endoribonuclease IRE1; serine/threonine-protein kinase/endoribonuclease IRE1-like
Gene Symbols ERN1
Host Species Rabbit
Immunogen Recombinant Human ERN1/IRE1 protein (Pro465-Leu977).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2081
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.