Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IREB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541782
Description
Peptide sequence: IQINLNSIVP SVSGPKRPQD RVAVTDMKSD FQACLNEKVG FKGFQIAAEK Sequence homology: Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 93%; Rat: 100%; Zebrafish: 93%.
Iron regulatory protein 2 (IRP2) is a central post-transcriptional regulator of cellular and systemic iron metabolism. It is a cytosolic RNA-binding protein that binds to stem-loop structures. IRP2 is regulated by iron-mediated proteolysis. Hypoxia is important for normal tissue physiology as well as being a component of several pathophysiological conditions, including heart and cerebrovascular diseases and tumor growth. Because iron and oxygen are closely related, IRP2 also plays a role during hypoxia (low oxygen).
Specifications
| IREB2 | |
| Polyclonal | |
| Unconjugated | |
| IREB2 | |
| ACO3; D9Ertd85e; FLJ23381; im:7153062; Ireb2; IRE-BP 2; IREBP2; Iron regulatory protein 2; iron responsive element binding protein 2; iron-regulatory protein 2; iron-responsive element binding protein 2; iron-responsive element-binding protein 2; Iron-responsive element-binding protein 2-like protein; IRP2; IRP2AD | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 3658, 64602 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P48200, Q811J3 | |
| IREB2 | |
| Synthetic peptide directed towards the middle region of human IREB2. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction