Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IREM-1 (CD300f) Mouse, anti-Human, R718, Clone: UP-D1, BD OptiBuild™

Catalog No. BDB752022 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB752022 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB752022 Supplier BD Biosciences Supplier No. 752022
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The UP-D1 monoclonal antibody specifically binds to Immune receptor expressed on myeloid cells 1 (IREM-1). IREM-1 is encoded by CD300LF. It is also known as CD300 antigen-like family member F (CD300f), CMRF35-like molecule 1 (CLM-1/CLM1), NK inhibitory receptor (NKIR), or Immunoglobulin superfamily member 13 (IgSF13). IREM-1/CD300f is expressed on monocytes, dendritic cells, granulocytes, and mast cells. IREM-1 is a single-pass type I transmembrane glycoprotein that belongs to the CMRF family of the Ig gene superfamily. The long intracellular region of IREM-1 contains two classical immunoreceptor tyrosine-based inhibitory motifs (ITIM). Ligand-bound IREM-1 can transduce negative regulatory signals intracellularly by recruiting SHP1 or SHIP phosphatases that in turn dephosphorylate tyrosine residues within activating cell signaling molecules. CD300f may thus play a role in regulating inflammatory responses involving myeloid cells.

TRUSTED_SUSTAINABILITY

Specifications

Antigen IREM-1 (CD300f)
Applications Flow Cytometry
Classification Monoclonal
Clone UP-D1
Conjugate R718
Host Species Mouse
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Refrigerate Upon Arrival
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.