Learn More
Description
Specifications
Specifications
| Antigen | IRF-1 |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | Please refer to lot specific datasheet. |
| Conjugate | Unconjugated |
| Gene Symbols | IRF1 |
| Host Species | Rabbit |
| Immunogen | KLH-conjugated linear peptide corresponding to 14 amino acids from the N-terminal half of human Interferon Regulatory Factor 1. |
| Purification Method | Affinity Purified |
| Quantity | 25 μg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.