Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF-2 Polyclonal Antibody, Active Motif™
SDP

Rabbit Polyclonal Antibody

$159.90 - $633.45

Specifications

Antigen IRF-2
Concentration Lot-Specific
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
50-199-3298
SDP
Encompass
View Documents
Active Motif
61282
10 μL
Each for $159.90
Only null left
Add to Cart
 
50-199-3297
SDP
Encompass
View Documents
Active Motif
61281
100 μL
Each for $633.45
Only null left
Add to Cart
 
Description

Description

IRF2 belongs to the interferon regulatory factor family. IRF2 regulates the transcription regulation important to cell proliferation as well as immune responses by binding to the upstream regulatory region of type I IFN and IFNinducible MHC class I genes (the interferon consensus sequence (ICS)) and repression of those genes. However, IRF2 also functions as a transcriptional activator of histone H4 by interactions with NF-kappa B.
Specifications

Specifications

IRF-2
Western Blot
Unconjugated
Rabbit
Human
P14316
3660
This IRF-2 antibody was raised against a peptide within the C-terminal region of human IRF-2.
Primary
-20°C, Avoid Freeze/Thaw Cycles
IRF2
Lot-Specific
Polyclonal
Liquid
RUO
PBS with 30% glycerol and 0.035% sodium azide; pH 7.5
9830146E22Rik; AI646973; DKFZp686F0244; Interferon regulatory factor 2; IRF2; IRF-2; OTTHUMP00000218576; OTTHUMP00000218580
IRF2
IgG
Antigen affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.