Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF-3 Mouse anti-Human, Alexa Fluor 647, Clone: SL-12.1 (also known as SL-12), BD

Catalog No. BDB566347 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566347 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566347 Supplier BD Biosciences Supplier No. 566347
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The SL-12.1 monoclonal antibody specifically recognizes human Interferon regulatory factor 3 (IRF-3). Viral infection in mammals can lead to the induction of multiple pathways as part of the host defense mechanism. One of the major pathways activated is the JAK-STAT pathway by various interferons (IFNαand IFNβ). These IFNs exert their influence via transcriptional activation of specific target genes involved in antiviral defense, for example the chemokine ISG15 gene or the major histocompatibility complex class I and II molecules. These genes in turn are regulated by the JAK-STAT signaling pathway and through interferon regulatory factors (IRFs). IRFs are a family of transcription factors that possess a broad range of activities. IRF-3 is one of nine members which all share a common DNA binding domain which binds to an IFN stimulated response element (ISRE) found in the majority of IFN-inducible promoters. The IRF3 /gene expresses a 50 kDa protein which is constitutively expressed in all tissues. The protein undergoes post-translational modification as well as dimerization and is translocated from the cytoplasm to the nucleus upon viral infection or exposure to dsRNA.

Alexa Fluor™ 647 conjugates are highly photostable and remain fluorescent over a broad pH range. The excitation and emission maxima are nearly identical to those of APC. However, APC tends to be brighter while Alexa Fluor™ 647 is more optimal for intracellular applications. This fluorochrome exhibits uncommon photostability, making it an ideal choice for use in fluorescence microscopy. Due to nearly identical excitation and emission properties but different spillover characteristics, APC and Alexa Fluor™ 647 cannot be used simultaneously.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF-3
Classification Monoclonal
Clone SL-12.1 (also known as SL-12)
Concentration 0.2mg/mL
Conjugate Alexa Fluor 647
Formulation Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide.
Gene Alias Interferon regulatory factor 3, IRF-3, IRF3, IIAE7
Host Species Mouse
Immunogen Human IRF-3 aa. 56-427 Recombinant Fusion Protein
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.