Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
IRF1 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP178763JF549
Description
IRF1 Polyclonal antibody specifically detects IRF1 in Mouse samples. It is validated for Western BlotSpecifications
IRF1 | |
Polyclonal | |
Western Blot | |
interferon regulatory factor 1, MARIRF-1 | |
The immunogen for this product maps to a region between residue 300 and 329 of mouse Interferon Regulatory Factor 1 using the numbering given in entry NP_032416.1 (GeneID 16362). | |
0.1 mL | |
Cell Cycle and Replication | |
3659 | |
Store at 4C in the dark. | |
IgG |
Western Blot | |
Janelia Fluor 549 | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction