Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

IRF1 Mouse anti-Human, PE, Clone: 20/IRF-1, BD

Catalog No. BDB566322 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB566322 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB566322 Supplier BD Biosciences Supplier No. 566322
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The 20/IRF-1 monoclonal antibody specifically binds to Interferon regulatory factor 1 (IRF1). IRF1 belongs to the human IRF family of transcription factors that contains nine members, IRF1-9. IRF1 is broadly expressed and its levels are upregulated in response to certain cytokines, interferons, bacterial or viral infections. IRF1 binds to specific DNA motifs such as IFN-stimulated response elements (ISRE) and regulates the activity of IFN-responsive genes. IRF1 plays roles in inflammation as well as innate and adaptive immune responses. It is involved in the regulation of cellular proliferation, Major Histocompatibility Complex (MHC) antigen expression, and the differentiation and effector functions of macrophages, dendritic cells, natural killer (NK) cells, and various T cell subsets. In addition, IRF1 may function as a tumor suppressor, since deletions in the IRF1 locus is associated with myelodysplasia and leukemias.

R-phycoerythrin (PE) is an accessory photosynthetic pigment found in red algae. It exists in vitro as a 240-kDa protein with 23 phycoerythrobilin chromophores per molecule. This makes PE one of the brightest fluorochromes for flow cytometry applications, but its photobleaching properties make it unsuitable for fluorescence microscopy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen IRF1
Classification Monoclonal
Clone 20/IRF-1
Concentration 0.2mg/mL
Conjugate PE
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Interferon regulatory factor 1, IRF-1, MAR
Host Species Mouse
Immunogen Human IRF1 aa. 159-279
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.