Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ IRF1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA550512
Description
The antibody detects endogenous levels of total IRF1 protein.
IRF1 encodes interferon regulatory factor 1, a member of the interferon regulatory transcription factor (IRF) family. IRF1 serves as an activator of interferons alpha and beta transcription, and in mouse it has been shown to be required for double-stranded RNA induction of these genes. IRF1 also functions as a transcription activator of genes induced by interferons alpha, beta, and gamma. Further, IRF1 has been shown to play roles in regulating apoptosis and tumor-suppression. Diseases associated with IRF1 include gastric cancer, somatic and lung cancer.
Specifications
IRF1 | |
Polyclonal | |
Unconjugated | |
IRF1 | |
AU020929; Interferon regulatory factor 1; interferon regulatory factor 1 isoform +I9; interferon regulatory factor 1 isoform d78; interferon regulatory factor 1 isoform delta4; interferon regulatory factor 1 isoform delta7; IRF1; IRF-1; MAR | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
16362, 3659 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1.5 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
P10914, P15314 | |
IRF1 | |
Synthetic peptide corresponding to a region derived from internal residues of human interferon regulatory factor 1. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction